Learn More
AdipoGen VEGF 164 mouse rec
SDP
Supplier:  AdipoGen AG40T0044C005
Encompass_Preferred
Protein. Mouse VEGF 164 164aa. Source Sf9 cells. Lyophilized. Purity >95% SDS-PAGE. Mouse vascular endothelial growth factor 164 VEGF 164 is produced as a homodimer. It binds to the FLT1/VEGFR1 and KDR/VEGFR2 receptors, heparan sulfate and heparin. It is a specific mitogen for vascular endothelial cells and a strong angiogenic factor in vivo. In addition to its action as a mitogen it is a potent vascular permeability factor VPF in vivo and is also a chemoattractant for monocytes and endothelial cells. It induces endothelial cell proliferation, promotes cell migration, inhibits apoptosis and induces permeabilization of blood vessels. It may play a role in increasing vascular permeability during lactation, when increased transport of molecules from the blood is required for efficient milk protein synthesis.
Catalog No. 50-256-2784