Learn More
AdipoGen VEGF 165 human rec
SDP
Supplier:  AdipoGen AG40T0045C020
Encompass_Preferred
Protein. Human VEGF 165 165aa. Source Sf9 cells. Lyophilized. Purity >95% SDS-PAGE. Human vascular endothelial growth factor VEGF 165 is produced as a homodimer. It binds to the FLT1/VEGFR1 and KDR/VEGFR2 receptors, heparan sulfate and heparin. VEGF 165 is a specific mitogen for vascular endothelial cells and a strong angiogenic factor in vivo. In addition to its action as a mitogen it is a potent vascular permeability factor VPF in vivo. It is also a chemoattractant molecule for monocytes and endothelial cells. It induces endothelial cell proliferation, promotes cell migration, inhibits apoptosis and induces permeabilization of blood vessels.
Catalog No. 50-256-2781