Learn More
AdipoGen VEGF-A h mAb 36D3
SDP
Supplier:  AdipoGen AG20T0105C200
Encompass_Preferred
Monoclonal Antibody. Isotype Mouse IgG1. Clone 36D3. Applications ELISA, FUNC, WB. Lyophilized. Human vascular endothelial erowth factor VEGF 165 is produced as a homodimer. It binds to the FLT1/VEGFR1 and KDR/VEGFR2 receptors, heparan sulfate and heparin. VEGF-A is a specific mitogen for vascular endothelial cells and a strong angiogenic factor in vivo. In addition to its action as a mitogen it is a potent vascular permeability factor VPF in vivo. It is also a chemoattractant molecule for monocytes and endothelial cells. It induces endothelial cell proliferation, promotes cell migration, inhibits apoptosis and induces permeabilization of blood vessels.
Catalog No. 50-256-3354