Learn More
Medchemexpress LLC VEGF-A Pig His 100ug
SDP
Catalog No. 5000182012
Encompass_Preferred
The VEGF-A protein is a multifunctional growth factor that is essential for promoting angiogenesis vasculogenesis and endothelial cell growth Its multiple effects include inducing endothelial cell proliferation promoting migration inhibiting apoptosis and increasing vascular permeability VEGF-A Protein Pig (His) is the recombinant pig-derived VEGF-A protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-82012 Supplier Medchemexpress LLC Supplier No. HYP72280100UG
The VEGF-A protein is a multifunctional growth factor that is essential for promoting angiogenesis vasculogenesis and endothelial cell growth Its multiple effects include inducing endothelial cell proliferation promoting migration inhibiting apoptosis and increasing vascular permeability VEGF-A Protein Pig (His) is the recombinant pig-derived VEGF-A protein expressed by E coli with N-6 His labeled tag