Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vascular endothelial growth factor A (VEGF-A), rabbit, His-tagged | >90.0% | 100UG
SDP

Catalog No. 5000176507
Encompass_Preferred

Recombinant rabbit vascular endothelial growth factor A (VEGF-A) is produced in Pichia pastoris with a 6xHis tag and supplied as a lyophilized powder for research applications in angiogenesis and endothelial biology.

  • Expressed in Pichia pastoris for eukaryotic post-translational modifications.
  • Includes N-terminal 6xHis tag for affinity purification or detection.
  • Observed molecular weight approximately 45-66 kDa; predicted mass 51.7 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for stability; store at -20°C.
  • Suitable for studies of angiogenesis, endothelial proliferation, migration, and permeability.

Catalog No. 50-001-76507 Supplier Medchemexpress LLC Supplier No. HYP700497100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rabbit vascular endothelial growth factor A (VEGF-A) is produced in Pichia pastoris with a 6xHis tag and supplied as a lyophilized powder for research applications in angiogenesis and endothelial biology.

  • Expressed in Pichia pastoris for eukaryotic post-translational modifications.
  • Includes N-terminal 6xHis tag for affinity purification or detection.
  • Observed molecular weight approximately 45-66 kDa; predicted mass 51.7 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for stability; store at -20°C.
  • Suitable for studies of angiogenesis, endothelial proliferation, migration, and permeability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.