Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vascular endothelial growth factor A (VEGF-A), recombinant rabbit, His-tag | >90.0% | 20UG
SDP

Catalog No. 5000175461
Encompass_Preferred

Recombinant rabbit VEGF-A (His-tagged) produced in Pichia pastoris, supplied as a lyophilized powder formulated with trehalose. The product contains residues Y51-R511 and is intended for research applications in angiogenesis and endothelial cell biology; follow supplier storage and reconstitution recommendations for optimal activity.

  • Recombinant His-tagged protein expressed in Pichia pastoris.
  • Construct covers residues Y51-R511.
  • Predicted molecular mass 51.7 kDa; ~45-66 kDa by SDS-PAGE.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for stability.
  • Stable at -20°C; freeze aliquots for long-term storage.
  • Do not reconstitute below 100 μg/mL in ddH2O.

Catalog No. 50-001-75461 Supplier Medchemexpress LLC Supplier No. HYP70049720UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rabbit VEGF-A (His-tagged) produced in Pichia pastoris, supplied as a lyophilized powder formulated with trehalose. The product contains residues Y51-R511 and is intended for research applications in angiogenesis and endothelial cell biology; follow supplier storage and reconstitution recommendations for optimal activity.

  • Recombinant His-tagged protein expressed in Pichia pastoris.
  • Construct covers residues Y51-R511.
  • Predicted molecular mass 51.7 kDa; ~45-66 kDa by SDS-PAGE.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for stability.
  • Stable at -20°C; freeze aliquots for long-term storage.
  • Do not reconstitute below 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.