Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7422MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IF, IHC. Conjugate/Tag: Unconjugated. Clone: VEGF/1063. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Tumor cells in astrocytomas, breast or ovarian carcinomas. Cellular Staining: Cytoplasmic, Cell Surface, Extracellular (Secreted). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human VEGF189 recombinant protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 19-22kDa (reducing), 38kDa-44kDa (non-reducing).Chromosome: 6p12. Gene Symbol: VEGFA. Gene ID: 7422. Ensemble: ENSG00000112715. Uniprot: NP_001191313.
Catalog No. 50-188-5268