Learn More
Biomatik Corporation VEGF Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50370
Encompass_Preferred
VEGF Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Vascular Endothelial Growth Factor A. The VEGF Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 27~232aa. Applications: ELISA, IHC, IF, WB. Target Synonyms: Vascular Endothelial Growth Factor A. Accession Number: P15692. Gene ID: 7422. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4387