Learn More
Medchemexpress LLC HY-P7313 10ug Medchemexpress, VEGF-C (HEK293-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501934380
Encompass_Preferred
Medchemexpress, HY-P7313 10ug VEGF-C (HEK293-expressed), Human CAS: Recombinant Human VEGF-C (HEK293-expressed) functions in lymphangiogenesis, where it acts on lymphatic endothelial cells (LECs) primarily via its receptor VEGFR-3 promoting survival, growth and migration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4380 Supplier Medchemexpress LLC Supplier No. HYP731310UG
Medchemexpress, HY-P7313 10ug VEGF-C (HEK293-expressed), Human CAS: Recombinant Human VEGF-C (HEK293-expressed) functions in lymphangiogenesis, where it acts on lymphatic endothelial cells (LECs) primarily via its receptor VEGFR-3 promoting survival, growth and migration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.