Learn More
Medchemexpress LLC VEGF-C Human 196a. 10ug
SDP
Catalog No. 5000185034
Encompass_Preferred
Recombinant human VEGF-C protein (196 amino acids, F32-R227) produced in HEK293 cells with a C-terminal 6xHis tag. Provided as a purified research reagent for in vitro studies of angiogenesis, lymphangiogenesis, and VEGF receptor signaling. Store frozen and aliquot to minimize freeze-thaw.
  • Recombinant human sequence corresponding to amino acids F32-R227.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag enables purification and detection.
  • Purified protein suitable for biochemical and cell-based assays.
  • Stable when stored frozen; aliquot to avoid repeated freeze-thaw cycles.
Catalog No. 50-001-85034 Supplier Medchemexpress LLC Supplier No. HYP7050110UG
Recombinant human VEGF-C protein (196 amino acids, F32-R227) produced in HEK293 cells with a C-terminal 6xHis tag. Provided as a purified research reagent for in vitro studies of angiogenesis, lymphangiogenesis, and VEGF receptor signaling. Store frozen and aliquot to minimize freeze-thaw.
  • Recombinant human sequence corresponding to amino acids F32-R227.
  • Expressed in HEK293 cells to preserve mammalian post-translational modifications.
  • C-terminal 6xHis tag enables purification and detection.
  • Purified protein suitable for biochemical and cell-based assays.
  • Stable when stored frozen; aliquot to avoid repeated freeze-thaw cycles.