Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VEGF-C Human 196a. 50ug
SDP

Supplier:  Medchemexpress LLC HYP7050150UG

Encompass_Preferred

Recombinant human VEGF-C fragment (196 amino acids; residues F32-R227) expressed in HEK293 cells and supplied as a lyophilized C-terminal His-tagged protein for research applications in angiogenesis and lymphangiogenesis. The product is purified and formulated for stability and low endotoxin suitable for in vitro and biochemical assays.

  • Recombinant human fragment spanning residues F32-R227.
  • Produced in HEK293 mammalian expression system.
  • C-terminal 6xHis tag for detection and purification.
  • Lyophilized powder formulated for long-term storage.
  • Purity ≥95% as determined by PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for cellular assays.
  • Observed molecular weight ~25-30 kDa due to glycosylation.

Catalog No. 50-001-85033


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.