Learn More
Medchemexpress LLC VEGF-D Mouse HEK29 10ug
SDP
Supplier:  Medchemexpress LLC HYP7347410UG
Encompass_Preferred
VEGF-D Protein a versatile growth factor orchestrates angiogenesis lymphangiogenesis and endothelial cell growth It stimulates proliferation migration and influences vessel permeability VEGF-D binds VEGFR-3 triggering crucial signaling for vascular development Structurally VEGF-D is a non-covalent homodimer emphasizing its intricate role in vascular processes VEGF-D Protein Mouse (HEK293 Fc) is the recombinant mouse-derived VEGF-D protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-80179