Learn More
Enzo Life Sciences VEGF (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1560001
Encompass_Preferred
The VEGF (human), EIA kit is a sensitive colorimetric immunometric enzyme immunoassay kit with results in just 2.5 hours. Absorbance is read at 450 nm. The wide dynamic range is ideal for various matrices. Assay up to 39 samples in duplicate with each kit. Alternative name: Vascular endothelial growth factor. Assay Time: 2.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3847