Learn More
Sigma Aldrich Fine Chemicals Biosciences VEGf human Animal componen
Catalog No. 502905110
Encompass
VEGf (vascular endothelial growth factor) signals through the three receptors fms-like tyrosine kinase (flt-1) KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant human VEGf is a 38.2kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.
Catalog No. 50-290-5110 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP318210UG
VEGf (vascular endothelial growth factor) signals through the three receptors fms-like tyrosine kinase (flt-1) KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant human VEGf is a 38.2kDa disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains.