Learn More
Genscript Corporation VEGF R2/KDRBiotin mFc Chimer
Catalog No. 5000058887
Encompass
Tyrosine-protein kinase that acts as a cell-surface receptor for VEGFA VEGFB and PGF and plays an essential role in the development of embryonic vasculature the regulation of angiogenesis cell survival cell migration macrophage function chemotaxis and cancer cell invasion The tyrosine kinase receptor vascular endothelial growth factor receptor 2 (VEGFR2) is a key regulator of angiogenesis
Catalog No. 50-000-58887 Supplier Genscript Corporation Supplier No. Z03972500
Tyrosine-protein kinase that acts as a cell-surface receptor for VEGFA VEGFB and PGF and plays an essential role in the development of embryonic vasculature the regulation of angiogenesis cell survival cell migration macrophage function chemotaxis and cancer cell invasion The tyrosine kinase receptor vascular endothelial growth factor receptor 2 (VEGFR2) is a key regulator of angiogenesis