Learn More
Genscript Corporation VEGF R3/FLT4 hFc Chimera Huma
Catalog No. 5000057929
Encompass
Vascular endothelial growth factor (VEGF) and its receptors VEGF-R1 -R2 and -R3 play important roles in tumor angiogenesis and are associated with poor prognosis in several solid tumors VEGF-R1 -R2 and -R3 were highly expressed in CRC cells and stromal vessels VEGF-R1 strong positive staining correlated with shorter survival after CRC surgery
Catalog No. 50-000-57929 Supplier Genscript Corporation Supplier No. Z03968100
Vascular endothelial growth factor (VEGF) and its receptors VEGF-R1 -R2 and -R3 play important roles in tumor angiogenesis and are associated with poor prognosis in several solid tumors VEGF-R1 -R2 and -R3 were highly expressed in CRC cells and stromal vessels VEGF-R1 strong positive staining correlated with shorter survival after CRC surgery