Learn More
Enzo Life Sciences VEGF Recombinant monoclonal antibody (VG76e) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6600200
Encompass_Preferred
Clone: VG76e. Isotype: IgGκ. Immunogen: Recombinant human VEGF189 expressed in E. coli. Species reactivity: Human, Mouse Primate. Applications: ELISA, IHC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3184