Learn More
Enzo Life Sciences VEGF Recombinant monoclonal antibody (VG76e) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3830200
Encompass_Preferred
Clone: Bevacizumab. Isotype: IgGκ. Immunogen: Recombinant human VEGF (isoform 165). Species reactivity: Human. Specificity: Recognizes native and reduced human VEGF (isoform 165). Applications: ELISA, Flow Cytometry, IP, WB, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2910