Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse VEGF120 Protein, His,Avitag™
Supplier:  ACROBiosystems VE0M82Q2200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse VEGF120, His,Avitag (VE0-M82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accession # Q00731-3).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 17.3 kDa. The protein migrates as 22-24 kDa and 24-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized VEGFR1/R2 at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His,Avitag (Cat. No. VE0-M82Q2) with a linear range of 0.1-2 ng/mL (QC tested).;Immobilized Avastin at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His,Avitag (Cat. No. VE0-M82Q2) with a linear range of 0.125-2 μg/mL (Routinely tested).
Catalog No. 50-195-7261