Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse VEGF120 Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;VE0,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Catalog No. 501658308
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse VEGF120 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Mouse VEGF120, His Tag (VE0-M82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accession # Q00731-3).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 17.3 kDa. The protein migrates as 22-24 kDa and 24-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized VEGFR1/R2 at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His Tag (Cat. No. VE0-M82Q2) with a linear range of 0.06-2 ng/mL (QC tested).;Immobilized Avastin at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His Tag (Cat. No. VE0-M82Q2) with a linear range of 0.125-2 μg/mL (Routinely tested).
Catalog No. 50-165-8308 Supplier ACROBiosystems Supplier No. VE0M82Q225UG
Recombinant Protein;Biotinylated Mouse VEGF120 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Mouse VEGF120, His Tag (VE0-M82Q2) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accession # Q00731-3).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 17.3 kDa. The protein migrates as 22-24 kDa and 24-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized VEGFR1/R2 at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His Tag (Cat. No. VE0-M82Q2) with a linear range of 0.06-2 ng/mL (QC tested).;Immobilized Avastin at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His Tag (Cat. No. VE0-M82Q2) with a linear range of 0.125-2 μg/mL (Routinely tested).