Learn More
Medchemexpress LLC VEGF121 Human HEK2 10ug
Supplier: Medchemexpress LLC HYP7051910UG
Recombinant human VEGF121 produced in HEK293 cells and supplied lyophilized for research applications. The protein demonstrates receptor-binding activity and stimulates endothelial cell proliferation, with bioactivity in the low ng/mL range, and is provided in a stability-optimized formulation.
- Recombinant human protein expressed in HEK293 cells.
- C-terminal His tag for detection and purification.
- Lyophilized from PBS with stabilizers for enhanced shelf life.
- High purity (>95%) by reducing SDS-PAGE.
- Demonstrated activity in receptor-binding and HUVEC proliferation assays (ED50 in low ng/mL).
- Store frozen and aliquot to minimize freeze-thaw cycles.