Learn More
Medchemexpress LLC VEGF121 Human HEK2 10ug
SDP
Catalog No. 5000180723
Encompass_Preferred
VEGF121 Protein is a subtype of Vascular Endothelial Growth Factor A (VEGF-A) VEGF-A is a key member of the VEGF family of cytokines that can promote neovascularization and increase vascular permeability VEGF121 Protein Human (HEK293) is the recombinant human-derived VEGF121 protein expressed by HEK293 with tag free The total length of VEGF121 Protein Human (HEK293) is 147 a a with molecular weight of 19 9 17 0 kDa respectively
Catalog No. 50-001-80723 Supplier Medchemexpress LLC Supplier No. HYP7347110UG
VEGF121 Protein is a subtype of Vascular Endothelial Growth Factor A (VEGF-A) VEGF-A is a key member of the VEGF family of cytokines that can promote neovascularization and increase vascular permeability VEGF121 Protein Human (HEK293) is the recombinant human-derived VEGF121 protein expressed by HEK293 with tag free The total length of VEGF121 Protein Human (HEK293) is 147 a a with molecular weight of 19 9 17 0 kDa respectively