Learn More
ACROBiosystems ACROBiosystems ActiveMax Human VEGF121 Protein, Tag Free
Supplier: ACROBiosystems VE1H421320UG
Recombinant Protein;20UG;ActiveMax Human VEGF121, Tag Free (VE1-H4213) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # P15692-9).;This protein carries no "tag". The protein has a calculated MW of 14.1 kDa. As a result of glycosylation, the protein migrates as 19 kDa and 20 kDa under reducing (R) condition, and 40-45 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human VEGF121, Tag Free (Cat. No. VE1-H4213) at 2 μg/mL (100 μL/well) can bind Human VEGF R2, Strep Tag (Cat. No. KDR-H5280) with a linear range of 0.15-2.5 μg/mL (QC tested).