Learn More
ACROBiosystems Recombinant Protein;Human VEGF121 Protein, His Tag;HEK293;1MG;VE1,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Catalog No. 501658808
Encompass_Preferred
Recombinant Protein;Human VEGF121 Protein, His Tag;1MG;Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # P15692-9).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15 kDa. As a result of glycosylation, the protein migrates as 17 kDa and 20 kDa under reducing (R) condition, and 35-45 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized VEGFR1/R2-Fc at 10 μg/mL (100 μL/well) can bind Human VEGF121, His Tag (Cat. No. VE1-H5246) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-8808 Supplier ACROBiosystems Supplier No. VE1H52461MG
Recombinant Protein;Human VEGF121 Protein, His Tag;1MG;Human VEGF121, His Tag (VE1-H5246) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 147 (Accession # P15692-9).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15 kDa. As a result of glycosylation, the protein migrates as 17 kDa and 20 kDa under reducing (R) condition, and 35-45 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized VEGFR1/R2-Fc at 10 μg/mL (100 μL/well) can bind Human VEGF121, His Tag (Cat. No. VE1-H5246) with a linear range of 0.4-13 ng/mL (QC tested).