Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse VEGF164 Protein, His,Avitag™
Catalog No. 501957391
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse VEGF164, His,Avitag (VE4-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 190 (Accession # Q00731-2).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 22.5 kDa. As a result of different glycosylation, the protein migrates as 28-33 kDa under reducing (R) condition, and 46-60 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized VEGFR1/R2, Fc Tag at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF164, His,Avitag (Cat. No. VE4-M82Q3) with a linear range of 0.02-0.5 ng/mL (QC tested).;Immobilized Human VEGF R1, His Tag (Cat. No. VE1-H5220) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF164, His,Avitag (Cat. No. VE4-M82Q3) with a linear range of 0.2-2 ng/mL (Routinely tested).
Catalog No. 50-195-7391 Supplier ACROBiosystems Supplier No. VE4M82Q325UG
Recombinant Protein;25UG;Biotinylated Mouse VEGF164, His,Avitag (VE4-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 190 (Accession # Q00731-2).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 22.5 kDa. As a result of different glycosylation, the protein migrates as 28-33 kDa under reducing (R) condition, and 46-60 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized VEGFR1/R2, Fc Tag at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF164, His,Avitag (Cat. No. VE4-M82Q3) with a linear range of 0.02-0.5 ng/mL (QC tested).;Immobilized Human VEGF R1, His Tag (Cat. No. VE1-H5220) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF164, His,Avitag (Cat. No. VE4-M82Q3) with a linear range of 0.2-2 ng/mL (Routinely tested).