Learn More
Medchemexpress LLC VEGF164 Mouse 164a 10ug
SDP
Catalog No. 5000184413
Encompass_Preferred
Recombinant mouse VEGF164 (164 amino acids) produced in Pichia pastoris and provided as a lyophilized protein for research applications involving angiogenesis and endothelial cell signaling. Supplied in a 10 μg package; consult the manufacturer datasheet for storage, reconstitution, and purity specifications.
  • Recombinant protein, 164 amino acids.
  • Expressed in Pichia pastoris expression system.
  • Lyophilized format for enhanced stability during storage.
  • Suitable for in vitro angiogenesis and endothelial cell assays.
  • High purity appropriate for research applications.
Catalog No. 50-001-84413 Supplier Medchemexpress LLC Supplier No. HYP7076110UG
Recombinant mouse VEGF164 (164 amino acids) produced in Pichia pastoris and provided as a lyophilized protein for research applications involving angiogenesis and endothelial cell signaling. Supplied in a 10 μg package; consult the manufacturer datasheet for storage, reconstitution, and purity specifications.
  • Recombinant protein, 164 amino acids.
  • Expressed in Pichia pastoris expression system.
  • Lyophilized format for enhanced stability during storage.
  • Suitable for in vitro angiogenesis and endothelial cell assays.
  • High purity appropriate for research applications.