Learn More
Medchemexpress LLC HY-K0221 30 mL Medchemexpress, Affinity Chromatography Column CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYK022130 ML5 PCS
Encompass_Preferred
Medchemexpress, HY-P7421 10ug VEGF164 (P.pastoris-expressed), Mouse CAS: Recombinant Mouse VEGF164 (P.pastoris-expressed), a VEGF isoform splice variant, is more potent at inducing endothelial ICAM-1 expression by VEGFR-2, leukocyte migration by VEGFR-1, and inflammation and angiogenesis in the adult cornea. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0208