Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP521410UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Mouse. BioActivity: HUVEC proliferation was significantly induced by less than 10 ng/ml, or activity of at least 1.0 x 10^5 units/mg. Typical ED50 values of new lots are generally lower, around 5 ng/ml. AA Sequence: MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPENHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR. Buffer: Sterile filtered (0.22 um) solution containing 0.1% TFA. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: VEGFA. Uniprot: Q00731-2.
Catalog No. 50-188-5715