Recombinant rat VEGF164 (isoform 164) expressed in P. pastoris and supplied lyophilized. The tag-free protein carries an A36T substitution, is formulated from 0.22 μm filtered PBS (pH 7.4), and is provided with >95% purity by reducing SDS-PAGE. It is intended for research applications in angiogenesis, endothelial cell signaling, and receptor binding assays; reported binding to VEGFR-1 demonstrates high affinity. Store frozen and aliquot to preserve activity.
- Recombinant rat VEGF164 isoform expressed in P. pastoris.
- Tag-free with A36T substitution.
- Lyophilized from 0.22 μm filtered PBS, pH 7.4.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Apparent molecular weight approximately 18-23 kDa under reducing conditions.
- Suitable for angiogenesis, endothelial cell, and receptor binding studies.
- Stable when stored frozen; aliquot to avoid repeated freeze-thaw cycles.