Learn More
ACROBiosystems ACROBiosystems ActiveMax Mouse VEGF164 Protein, Tag Free (HPLC-verified)
Supplier:  ACROBiosystems VE4M421620UG
Encompass_Preferred
Recombinant Protein;20UG;ActiveMax Mouse VEGF164, Tag Free (HPLC-verified) (VE4-M4216) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 190 (Accession # AAB22253.1 ).;This protein carries no "tag". The protein has a calculated MW of 19.3 kDa. The protein migrates as 23-30 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized ActiveMax Mouse VEGF164, Tag Free (HPLC-verified) (Cat. No. VE4-M4216) at 2 μg/mL (100 μL/well) can bind VEGFR1/R2-Fc with a linear range of 0.6-2.5 ng/mL (QC tested).
Catalog No. 50-195-7548