Learn More
ACROBiosystems ACROBiosystems Biotinylated Human VEGF165 Protein, His,Avitag™
Supplier:  ACROBiosystems VE5H82Q0200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human VEGF165, His,Avitag (VE5-H82Q0) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 22.4 kDa (monomer). As a result of glycosylation, the protein migrates as 30 kDa (monomer) under reducing (R) condition, and 48-60 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human VEGFR1/R2, Fc tag at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF165, His,Avitag (Cat. No. VE5-H82Q0) with a linear range of 0.5-7.8 ng/mL (QC tested).
Catalog No. 50-195-7408