Learn More
Medchemexpress LLC HY-P7312 10ug Medchemexpress, VEGF165, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934379
Encompass_Preferred
Medchemexpress, HY-P7312 10ug VEGF165, Mouse CAS: Recombinant Mouse VEGF165 is a potent mediator of angiogenesis and binds the semaphorin receptor, neuropilin1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4379 Supplier Medchemexpress LLC Supplier No. HYP731210UG
Medchemexpress, HY-P7312 10ug VEGF165, Mouse CAS: Recombinant Mouse VEGF165 is a potent mediator of angiogenesis and binds the semaphorin receptor, neuropilin1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.