Learn More
ACROBiosystems Recombinant Protein;Human VEGF165 Protein, His Tag;HEK293;1MG;VE5,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Catalog No. 501658165
Encompass_Preferred
Recombinant Protein;Human VEGF165 Protein, His Tag;1MG;Human VEGF165, His Tag (VE5-H5248) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 24 kDa and 25-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF R1&VEGF R2, Fc Tag at 2 μg/mL (100 μL/well) can bind Human VEGF165, His Tag (Cat. No. VE5-H5248) with a linear range of 0.2-4 ng/mL (QC tested).;Avastin (Bevacizumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human VEGF165 Protein, His Tag (Cat. No. VE5-H5248) with an affinity constant of 1.03 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-165-8165 Supplier ACROBiosystems Supplier No. VE5H52481MG
Recombinant Protein;Human VEGF165 Protein, His Tag;1MG;Human VEGF165, His Tag (VE5-H5248) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 24 kDa and 25-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF R1&VEGF R2, Fc Tag at 2 μg/mL (100 μL/well) can bind Human VEGF165, His Tag (Cat. No. VE5-H5248) with a linear range of 0.2-4 ng/mL (QC tested).;Avastin (Bevacizumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human VEGF165 Protein, His Tag (Cat. No. VE5-H5248) with an affinity constant of 1.03 nM as determined in a SPR assay (Biacore T200) (Routinely tested).