Learn More
Aviva Systems Biology Corp VEGFA Antibody - C-terminal region (ARP60981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VEGFA. It was validated on Western Blot. . Alternate names: MGC70609, MVCD1, VEGF, VPFApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-287-221