Learn More
Sino Biological Anti-IL-3R Beta/IL3RB Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11066MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD131 / CSF2RB / IL3RB / IL5RB (rh CD131 / CSF2RB / IL3RB / IL5RB; Catalog#10516-H08H; NP_000386.1; Met1-Trp443). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8596