Learn More
Aviva Systems Biology Corp VEGFB Antibody - middle region (ARP58975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VEGFB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human VEGFB. Alternate names: VEGFL, VRFApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-547