Learn More
Aviva Systems Biology Corp VEGFB Antibody - C-terminal region (ARP58976_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58976_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VEGFB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human VEGFB. Alternate names: VEGFL, VRFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-548