Learn More
Sino Biological Anti-VEGF-C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10542T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VEGFC/VEGF-C/Flt4-L (rh VEGFC/VEGF-C/Flt4-L; Catalog#10542-H08H; NP_005420.1; Thr103-Arg227). VEGFC/VEGF-C/Flt4-L specific IgG was purified by Human VEGFC/VEGF-C/Flt4-L affinity chromatography.
Catalog No. 50-224-7835