Learn More
Medchemexpress LLC VEGFR-1 Rat HEK293 2ug
SDP
Catalog No. 5000180056
Encompass_Preferred
VEGFR-1 protein is a tyrosine protein kinase receptor for VEGFA VEGFB and PGF and is critical for embryonic vasculature development angiogenesis cell survival migration macrophage function chemotaxis and cancer invasion It actively regulates postnatal retinal vitreous vascular degeneration and may act as a negative regulator of embryonic angiogenesis VEGFR-1 Protein Rat (HEK293 His) is the recombinant rat-derived VEGFR-1 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80056 Supplier Medchemexpress LLC Supplier No. HYP735522UG
VEGFR-1 protein is a tyrosine protein kinase receptor for VEGFA VEGFB and PGF and is critical for embryonic vasculature development angiogenesis cell survival migration macrophage function chemotaxis and cancer invasion It actively regulates postnatal retinal vitreous vascular degeneration and may act as a negative regulator of embryonic angiogenesis VEGFR-1 Protein Rat (HEK293 His) is the recombinant rat-derived VEGFR-1 protein expressed by HEK293 with C-His labeled tag