Learn More
Enzo Life Sciences VEGFR 2 (human), (recombinant) (GST-tag) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4040010
Encompass_Preferred
Alternative name: KDR, FLK 1, Vascular endothelial growth factor receptor 2. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 40mM TRIS-HCl, pH 8.0, 110mM sodium chloride, 2.2mM potassium chloride, 20% Glycerol, and 3mM DTT. Source: Produced in Sf9 insect cells with N-terminal GST tage. Human VEGFR2 (aa 805-1356). Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2010