Learn More
Medchemexpress LLC VEGFR-2-IN-5 hydrochloride | 1430089-64-7 | 364.45 | 5 MG
SDP
Catalog No. 5000388012
Encompass_Preferred
VEGFR-2-IN-5 is a VEGFR2 inhibitor extracted from patent WO2013055780A1, Page 69. At equivalent molar concentrations, both the salt and free forms of a compound exhibit comparable biological activity. Nevertheless, the salt form (VEGFR-2-IN-5 hydrochloride) usually boasts enhanced water solubility and stability. It is for research use only and not sold to patients.
  • VEGFR2 inhibitor.
  • The salt form offers enhanced water solubility and stability compared to the free form.
  • For research use only.
Catalog No. 50-003-88012 Supplier Medchemexpress LLC Supplier No. HY18625A5MG
VEGFR-2-IN-5 is a VEGFR2 inhibitor extracted from patent WO2013055780A1, Page 69. At equivalent molar concentrations, both the salt and free forms of a compound exhibit comparable biological activity. Nevertheless, the salt form (VEGFR-2-IN-5 hydrochloride) usually boasts enhanced water solubility and stability. It is for research use only and not sold to patients.
  • VEGFR2 inhibitor.
  • The salt form offers enhanced water solubility and stability compared to the free form.
  • For research use only.