Learn More
AdipoGen VEGFR-2 KDR h mAb EIC
SDP
Supplier:  AdipoGen AG20T0108C100
Encompass_Preferred
Monoclonal Antibody. The antibody will detect native human VEGFR-2/KDR in ELISA experiments and on the surface of different human cell types. Isotype Mouse IgG1. Clone EIC. Applications ELISA, FACS, IHC, WB. Lyophilized. Disruption of the precise balance of positive and negative molecular regulators of blood and lymphatic vessel growth can lead to myriad diseases. Although dozens of natural inhibitors of hemangiogenesis have been identified, an endogenous selective inhibitor of lymphatic vessel growth has not been previously described. A splice variant of the gene encoding vascular endothelial growth factor receptor-2 VEGFR-2 that encodes a secreted form of the protein, designated endogenous soluble VEGFR-2 esVEGFR-2/KDR has been described. The endogenous soluble esKDR inhibits developmental and reparative lymphangiogenesis by blocking VEGF-C function.
Catalog No. 50-256-3351