Learn More
Enzo Life Sciences VEGFR 2 substrate (biotinylated) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2410001
Encompass_Preferred
This peptide, Biotin-Ahx-EEEEAYGWLDF, is derived from gastrin and can be used as a substrate for KDR/VEGFR2. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes. Alternative name: KDR substrate (biotinylated). Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1306