Learn More
TARGETMOL CHEMICALS INC VEGFR-3-IN-1 5MG
SDP
Catalog No. 502579229
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. VEGFR-3-IN-1 is a novel potent and selective VEGFR3 inhibitor with an IC50 of 110.4 nM.VEGFR-3-IN-1 possesses antitumor activity inactivates the VEGFR3 signaling pathway and inhibits the proliferation and migration of VEGF-C-induced human dermal lymphatic endothelial cells (HDLEC) MDA-MB-231 and MDA-MB-436 cells. MB-436 cells and inhibited the proliferation and migration of human dermal lymphatic endothelial cells (HDLEC) MDA-MB-231 and MDA-MB-436 cells. purity: 99%
Catalog No. 50-257-9229 Supplier TARGETMOL CHEMICALS INC Supplier No. T729245MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. VEGFR-3-IN-1 is a novel potent and selective VEGFR3 inhibitor with an IC50 of 110.4 nM.VEGFR-3-IN-1 possesses antitumor activity inactivates the VEGFR3 signaling pathway and inhibits the proliferation and migration of VEGF-C-induced human dermal lymphatic endothelial cells (HDLEC) MDA-MB-231 and MDA-MB-436 cells. MB-436 cells and inhibited the proliferation and migration of human dermal lymphatic endothelial cells (HDLEC) MDA-MB-231 and MDA-MB-436 cells. purity: 99%