Learn More
Selleck Chemical LLC Imatinib (STI571) 250mg 152459-95-5 Gleevec, Glivec, CGP057148B
SDP
Catalog No. NC0256071
Encompass_Preferred
Imatinib (STI571, CGP057148B, Gleevec) is a multi-target inhibitor of tyrosine kinase with inhibition for v-Abl, c-Kit and PDGFR, IC50 values are 0.6 ?M, 0.1 ?M and 0.1 ?M in cell-free or cell-based assays, respectively. Imatinib (STI571) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0256071 Supplier Selleck Chemical LLC Supplier No. S2475250MG
Add to Cart
Add to Cart
Imatinib (STI571, CGP057148B, Gleevec) is a multi-target inhibitor of tyrosine kinase with inhibition for v-Abl, c-Kit and PDGFR, IC50 values are 0.6 ?M, 0.1 ?M and 0.1 ?M in cell-free or cell-based assays, respectively. Imatinib (STI571) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.