Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human VEGF R1 / Flt-1 Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;VE1,FLT,VEGFR1,VEGFR-1,VEGF R1,FLT1,FLT-1
Catalog No. 501658033
Encompass_Preferred
Recombinant Protein;Biotinylated Human VEGF R1 / Flt-1 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human VEGF R1, His Tag, Avi Tag (VE1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Asn 756 (Accession # P17948-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 85.9 kDa. The protein migrates as 106-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R1, His Tag, Avi Tag (Cat. No. VE1-H82E3) with a linear range of 1-31 ng/mL (QC tested).;Immobilized Human PLGF, His Tag (Cat. No. PGF-H5229) at 1 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R1, His Tag, Avi Tag (Cat. No. VE1-H82E3) with a linear range of 2-20 ng/mL (Routinely tested).
Catalog No. 50-165-8033 Supplier ACROBiosystems Supplier No. VE1H82E3200UG
Recombinant Protein;Biotinylated Human VEGF R1 / Flt-1 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human VEGF R1, His Tag, Avi Tag (VE1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Asn 756 (Accession # P17948-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 85.9 kDa. The protein migrates as 106-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R1, His Tag, Avi Tag (Cat. No. VE1-H82E3) with a linear range of 1-31 ng/mL (QC tested).;Immobilized Human PLGF, His Tag (Cat. No. PGF-H5229) at 1 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R1, His Tag, Avi Tag (Cat. No. VE1-H82E3) with a linear range of 2-20 ng/mL (Routinely tested).