Learn More
ACROBiosystems Biotinylated Human VEGF R2 Protein, Avitag™, His Tag, 200ug
Catalog No. 501957431
Encompass_Preferred
Vascular endothelial growth factor receptor 2 (VEGFR2) is a key receptor for vascular endothelial growth factor (VEGF) that mediates cellular responses to VEGF, playing a vital role in angiogenesis. This biotinylated recombinant human protein is expressed in human 293 cells (HEK293) and spans amino acids Ala 20 to Glu 764. It is tagged with an Avitag™ at the C-terminus followed by a polyhistidine tag, with biotinylation enzymatically performed on the single lysine residue within the Avitag.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 85.9 kDa, migrating at 120–150 kDa under reducing conditions due to glycosylation
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4, with trehalose as a protectant
  • Validated for use in ELISA, BLI, and FACS applications
  • Stable for 12 months at -20°C to -70°C in lyophilized state
Catalog No. 50-195-7431 Supplier ACROBiosystems Supplier No. KDRH82E5200UG
Vascular endothelial growth factor receptor 2 (VEGFR2) is a key receptor for vascular endothelial growth factor (VEGF) that mediates cellular responses to VEGF, playing a vital role in angiogenesis. This biotinylated recombinant human protein is expressed in human 293 cells (HEK293) and spans amino acids Ala 20 to Glu 764. It is tagged with an Avitag™ at the C-terminus followed by a polyhistidine tag, with biotinylation enzymatically performed on the single lysine residue within the Avitag.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 85.9 kDa, migrating at 120–150 kDa under reducing conditions due to glycosylation
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4, with trehalose as a protectant
  • Validated for use in ELISA, BLI, and FACS applications
  • Stable for 12 months at -20°C to -70°C in lyophilized state