Learn More
Medchemexpress LLC Vegfr2-in-2 | 737818-56-3 | 99.3% | C15H11BrN2O | 10MG
SDP
Catalog No. 5000258636
Encompass_Preferred
VEGFR2-IN-2 is a potent and selective VEGFR2 inhibitor (compound 6e) reported with an IC50 of 19.32 nM and intended for laboratory research into angiogenesis and oncology. The material is supplied as a solid with supporting SDS and COA documents available for verification.
  • Potent VEGFR2 inhibitor with IC50 = 19.32 nM.
  • High reported purity of 99.3% (COA).
  • Solid; white to off-white appearance.
  • Molecular formula C15H11BrN2O, molecular weight 315.16.
  • Available in small-scale quantities (1 mg-100 mg) and as a 10 mM solution.
  • SDS and certificate of analysis available for verification.
  • Intended for in vitro research use only; not for human or veterinary use.
Catalog No. 50-002-58636 Supplier Medchemexpress LLC Supplier No. HY14713310MG
VEGFR2-IN-2 is a potent and selective VEGFR2 inhibitor (compound 6e) reported with an IC50 of 19.32 nM and intended for laboratory research into angiogenesis and oncology. The material is supplied as a solid with supporting SDS and COA documents available for verification.
  • Potent VEGFR2 inhibitor with IC50 = 19.32 nM.
  • High reported purity of 99.3% (COA).
  • Solid; white to off-white appearance.
  • Molecular formula C15H11BrN2O, molecular weight 315.16.
  • Available in small-scale quantities (1 mg-100 mg) and as a 10 mM solution.
  • SDS and certificate of analysis available for verification.
  • Intended for in vitro research use only; not for human or veterinary use.