Learn More
Cayman Chemical VEGFR2 Kinase Inhibitor I, Purity: ≥95%, CAS: 15966-93-5, 10 mg
SDP
Catalog No. 502955665
Encompass_Preferred
A potent, cell-permeable inhibitor of mouse VEGFR2 kinase (IC50 = 70 nM); has little or no effect against PDGFR, EGFR or IGF1R kinases (IC50 > 100 µM)
Catalog No. 50-295-5665 Supplier Cayman Chemical Supplier No. 1357810 MG
A potent, cell-permeable inhibitor of mouse VEGFR2 kinase (IC50 = 70 nM); has little or no effect against PDGFR, EGFR or IGF1R kinases (IC50 > 100 µM)