Learn More
Cayman Chemical VEGFR2 KInase InhIbItr I 5mg
SDP
Catalog No. 502955104
Encompass_Preferred
A potent, cell-permeable inhibitor of mouse VEGFR2 kinase (IC50 = 70 nM); has little or no effect against PDGFR, EGFR or IGF1R kinases (IC50 > 100 µM)
Catalog No. 50-295-5104 Supplier Cayman Chemical Supplier No. 135785 MG
Add to Cart
Add to Cart
A potent, cell-permeable inhibitor of mouse VEGFR2 kinase (IC50 = 70 nM); has little or no effect against PDGFR, EGFR or IGF1R kinases (IC50 > 100 µM)